Learn More
CPC Scientific H-Ile-Leu-Pro-Trp-Lys-Trp-Pro-Trp-Trp-Pro-Trp-Arg-Arg-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ATBP004B
Encompass
13-residue peptide amide isolated and characterized from the cytoplasmic granules of bovine neutrophils, having potent antibacterial activity. It is also highly cytotoxic to T-line cells. Furthermore, the primary structure of indolicidin is characterized by 5 tryptophan residues, which is the highest mole percentage of any known protein sequence and is unique among known endogenous antibacterial peptides. ONE-LETTER SEQUENCE: ILPWKWPWWPWRR-NH2MOLECULAR FORMULA: C100H132N26O13MOLECULAR WEIGHT:1906.3STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [140896-21-5], RESEARCH AREA: AntimicrobialREFERENCES: C. Subbalakshmi et al., FEBS Letters, 395, 48 (1996)
Catalog No. 50-210-8798