Learn More
Bachem Indolicidin
SDP
Supplier:  Bachem 4030762.1000
Encompass_Preferred
Indolicidin, H-Ile-Leu-Pro-Trp-Lys-Trp-Pro-Trp-Trp-Pro-Trp-Arg-Arg-NH2, 140896-21-5, C100H132N26O13, Mr 1906.32, 1mg. This cationic, tryptophan-rich peptide was purified from bovine granulocytes. Indolicidin shows potent antimicrobial activity in vitro against bacteria and fungi. It is also highly cytotoxic to T-line cells.
Catalog No. 50-259-478