Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indolicidin (acetate) | 99.6% | 1966.34 | 10 MG
SDP

Catalog No. 5000425071
Encompass_Preferred

Indolicidin acetate is a potent antimicrobial peptide purified from the cytoplasmic granules of bovine neutrophils. This product is for research use only.

  • Potent antimicrobial peptide
  • Purified from bovine neutrophils
  • Molecular formula: C102H136N26O15
  • Appearance: White to off-white solid
  • Purity: 99.6%
  • Molecular weight: 1966.34

Catalog No. 50-004-25071 Supplier Medchemexpress LLC Supplier No. HYP0261A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indolicidin acetate is a potent antimicrobial peptide purified from the cytoplasmic granules of bovine neutrophils. This product is for research use only.

  • Potent antimicrobial peptide
  • Purified from bovine neutrophils
  • Molecular formula: C102H136N26O15
  • Appearance: White to off-white solid
  • Purity: 99.6%
  • Molecular weight: 1966.34

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.