Learn More
Cayman Chemical Indolicidin trifluoroacetate
SDP
Catalog No. 502909260
Encompass_Preferred
An antimicrobial peptide; active against MDR-EAEC (MICs = 32 µM for all); active against S. cerevisiae, T. beigelii, and C. albicans (MICs = 5-10, 2.5-5, and 5-10 µM, respectively); reduces viral replication of HIV-1 in MT-2 cells from 67-100 UG/ml; increases survival of MDR-EAEC-infected G. mellonella larvae
Catalog No. 50-290-9260 Supplier Cayman Chemical Supplier No. 35845500 UG
An antimicrobial peptide; active against MDR-EAEC (MICs = 32 µM for all); active against S. cerevisiae, T. beigelii, and C. albicans (MICs = 5-10, 2.5-5, and 5-10 µM, respectively); reduces viral replication of HIV-1 in MT-2 cells from 67-100 UG/ml; increases survival of MDR-EAEC-infected G. mellonella larvae