Learn More
Cayman Chemical Indolicidin (trifluoroacetate salt), Purity: ≥98%, 5 mg
SDP
Catalog No. 502909231
Encompass_Preferred
An antimicrobial peptide; active against MDR-EAEC (MICs = 32 µM for all); active against S. cerevisiae, T. beigelii, and C. albicans (MICs = 5-10, 2.5-5, and 5-10 µM, respectively); reduces viral replication of HIV-1 in MT-2 cells from 67-100 µg/ml; increases survival of MDR-EAEC-infected G. mellonella larvae
Catalog No. 50-290-9231 Supplier Cayman Chemical Supplier No. 358455 MG
An antimicrobial peptide; active against MDR-EAEC (MICs = 32 µM for all); active against S. cerevisiae, T. beigelii, and C. albicans (MICs = 5-10, 2.5-5, and 5-10 µM, respectively); reduces viral replication of HIV-1 in MT-2 cells from 67-100 µg/ml; increases survival of MDR-EAEC-infected G. mellonella larvae