Learn More
Medchemexpress LLC Indoline | 496-15-1 | MFCD00005705 | 99.9% | 119.17 g/mol | C8H9N | 250g
SDP
Catalog No. 5000268604
Encompass_Preferred
Indoline is a derivative of Indole (HY-W001132) Indoline can use as the basic structure for CD4 mimetic compounds (CD4mcs) which triggers conformational changes of the HIV-1 envelope glycoprotein (Env) in advance and causes viral inactivation Indoline binds to gp120 Indoline CD4mcs can inhibit HIV-1AD8 with an IC50 in the micromolar range[1]
Catalog No. 50-002-68604 Supplier Medchemexpress LLC Supplier No. HYY0788250G
Indoline is a derivative of Indole (HY-W001132) Indoline can use as the basic structure for CD4 mimetic compounds (CD4mcs) which triggers conformational changes of the HIV-1 envelope glycoprotein (Env) in advance and causes viral inactivation Indoline binds to gp120 Indoline CD4mcs can inhibit HIV-1AD8 with an IC50 in the micromolar range[1]