Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indoline, 99%

Catalog No. p-7081152
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1100018 50 g
AAA1100030 250 g
AAA110000B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1100018 Supplier Thermo Scientific Chemicals Supplier No. A1100018
Only null left
Add to Cart

CAS: 496-15-1 | C8H9N | 119.167 g/mol

Indole is used in perfumery and in preparing tryptophan, one of the 20 amino acids commonly found in animal proteins. It has important application in the industry of plant growth. It is used to prepare indoleacetic acid (auxin) and other plant growth substances which help the development of roots in plant. Indole and its derivatives are widely used in making perfumes, dyes, agrochemicals and medicines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole is used in perfumery and in preparing tryptophan, one of the 20 amino acids commonly found in animal proteins. It has important application in the industry of plant growth. It is used to prepare indoleacetic acid (auxin) and other plant growth substances which help the development of roots in plant. Indole and its derivatives are widely used in making perfumes, dyes, agrochemicals and medicines.

Solubility
Fully misicible in water at 5 g/L (20°C).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents, acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-15-1
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.167
MDL Number MFCD00005705
InChI Key LPAGFVYQRIESJQ-UHFFFAOYSA-N
Synonym indoline, 2,3-dihydroindole, 1-azaindan, dihydroindole, 1h-indole, 2,3-dihydro, azaindane, benzopyrrolidine, aza-indane, indoline, 19, pubchem7502
PubChem CID 10328
ChEBI CHEBI:43295
IUPAC Name 2,3-dihydro-1H-indole
SMILES C1CNC2=CC=CC=C21

Specifications

Melting Point -21°C
Density 1.062
Boiling Point 220°C to 221°C
Flash Point 92°C (197°F)
Odor Odorless
Refractive Index 1.593
Quantity 50 g
Beilstein 111915
Sensitivity Light sensitive
Solubility Information Fully misicible in water at 5g/L (20°C).
Formula Weight 119.17
Percent Purity 99%
Chemical Name or Material Indoline
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 207-816-8
RTECSNumber NL6906300
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.