Learn More
Selleck Chemical LLC Indometacin Sodium S5010-1g
SDP
Catalog No. 503110169
Encompass_Preferred
Indometacin Sodium (Indomethacin Sodium) is the sodium salt of indomethacin which is a non-selective reversible and competitive inhibitor of cyclooxygenases 1 and 2 with anti-inflammatory analgesic-antipyretic and tocolytic effects
Catalog No. 50-311-0169 Supplier Selleck Chemical LLC Supplier No. S50101G
Indometacin Sodium (Indomethacin Sodium) is the sodium salt of indomethacin which is a non-selective reversible and competitive inhibitor of cyclooxygenases 1 and 2 with anti-inflammatory analgesic-antipyretic and tocolytic effects