Learn More
Selleck Chemical LLC Indometacin Sodium S5010-25mg
SDP
Catalog No. 503111977
Encompass_Preferred
Indometacin Sodium (Indomethacin Sodium) is the sodium salt of indomethacin which is a non-selective reversible and competitive inhibitor of cyclooxygenases 1 and 2 with anti-inflammatory analgesic-antipyretic and tocolytic effects
Catalog No. 50-311-1977 Supplier Selleck Chemical LLC Supplier No. S501025MG
Indometacin Sodium (Indomethacin Sodium) is the sodium salt of indomethacin which is a non-selective reversible and competitive inhibitor of cyclooxygenases 1 and 2 with anti-inflammatory analgesic-antipyretic and tocolytic effects