Learn More
Apexbio Technology LLC Indomethacin(Synonyms: Indocin, Indometacin, Indomethacine, Indocid, Indocine, Amuno, Chrono-Indocid, Elmetacin), 10mM (in 1mL DMSO), CAS: 53-86-1.
SDP
Catalog No. 5000060376
Encompass_Preferred
Indomethacin (CAS 53-86-1) is a nonsteroidal anti-inflammatory compound that inhibits cyclooxygenase enzymes Cox-1 and Cox-2 displaying preferential inhibition toward Cox-1 (IC50 230 nM) compared to Cox-2 (IC50 630 nM) Additionally it acts as an agonist of peroxisome proliferator-activated receptor gamma (PPAR ) a transcriptional regulator implicated in adipogenesis Indomethacin can also activate PPAR Furthermore studies indicate that indomethacin stabilizes cholesterol-rich nanoscale clusters enhancing membrane phase separation and potentially altering membrane-dependent signaling pathways This profile makes indomethacin useful for investigating inflammation-associated processes lipid metabolism and membrane signaling mechanisms
Catalog No. 50-000-60376 Supplier Apexbio Technology LLC Supplier No. A84495.1
Indomethacin (CAS 53-86-1) is a nonsteroidal anti-inflammatory compound that inhibits cyclooxygenase enzymes Cox-1 and Cox-2 displaying preferential inhibition toward Cox-1 (IC50 230 nM) compared to Cox-2 (IC50 630 nM) Additionally it acts as an agonist of peroxisome proliferator-activated receptor gamma (PPAR ) a transcriptional regulator implicated in adipogenesis Indomethacin can also activate PPAR Furthermore studies indicate that indomethacin stabilizes cholesterol-rich nanoscale clusters enhancing membrane phase separation and potentially altering membrane-dependent signaling pathways This profile makes indomethacin useful for investigating inflammation-associated processes lipid metabolism and membrane signaling mechanisms