Learn More
Medchemexpress LLC HY-14397 1g Medchemexpress, Indomethacin CAS:53-86-1 Purity:>98%
SDP
Catalog No. 502029048
Encompass_Preferred
Medchemexpress, HY-14397 1g Indomethacin CAS:53-86-1 Indomethacin (Indometacin) is a potent, blood-brain permeable and nonselective inhibitor of COX1 and COX2, with IC50s of 18 nM and 26 nM for human COX-1 and COX-2, respectively, in CHO cells. Indomethacin disrupts autophagic flux by disturbing the normal functioning of lysosomes[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9048 Supplier Medchemexpress LLC Supplier No. HY143971G
Medchemexpress, HY-14397 1g Indomethacin CAS:53-86-1 Indomethacin (Indometacin) is a potent, blood-brain permeable and nonselective inhibitor of COX1 and COX2, with IC50s of 18 nM and 26 nM for human COX-1 and COX-2, respectively, in CHO cells. Indomethacin disrupts autophagic flux by disturbing the normal functioning of lysosomes[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.