Learn More
Cayman Chemical IndomethacIn 1g
SDP
Catalog No. 502896734
Encompass_Preferred
A non-selective COX inhibitor (IC50s = 1.67 and 24.6 µM for human COX-1 and COX-2, respectively); reduces filter paper-disc induced growth of granulation tissue in chick chorioallantoic membranes; reduces ocular inflammation induced by bovine serum in rabbits; reduces paw edema in a rat model of carrageenan-induced inflammation
Catalog No. 50-289-6734 Supplier Cayman Chemical Supplier No. 702701 G
A non-selective COX inhibitor (IC50s = 1.67 and 24.6 µM for human COX-1 and COX-2, respectively); reduces filter paper-disc induced growth of granulation tissue in chick chorioallantoic membranes; reduces ocular inflammation induced by bovine serum in rabbits; reduces paw edema in a rat model of carrageenan-induced inflammation