Learn More
Medchemexpress LLC HY-14397 500mg , Indomethacin CAS:53-86-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14397500MG
Encompass_Preferred
HY-14397 500mg Indomethacin CAS: 53-86-1 Indomethacin (Indometacin) is a potent, orally active COX1/2 inhibitor with IC50 values of 18 nM and 26 nM for COX-1 and COX-2, respectively. Indomethacin has anticancer activity and anti-infective activity. Indomethacin can be used for cancer, inflammation and viral infection research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9518