Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indomethacin | 53-86-1 | MFCD00057095 | 99.90% | C19H16ClNO4 | 1 ML
SDP

Catalog No. 5000355040
Encompass_Preferred

Indomethacin is a potent, orally active COX1/2 inhibitor, demonstrating significant anti-inflammatory, anticancer, and anti-infective activities. It is a widely used non-steroidal anti-inflammatory drug (NSAID) suitable for various research applications.

  • Potent, orally active COX1/2 inhibitor
  • Exhibits anticancer activity
  • Exhibits anti-infective activity
  • Suitable for research applications in cancer, inflammation, and viral infection
  • Inhibits both COX-1 (IC50 18 nM) and COX-2 (IC50 26 nM)
  • Provides anti-inflammatory and analgesic effects
  • Used to prevent preterm labor due to prostaglandin synthesis inhibition
  • Used in premature infants with patent ductus arteriosus

Catalog No. 50-003-55040 Supplier Medchemexpress LLC Supplier No. HY1439710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indomethacin is a potent, orally active COX1/2 inhibitor, demonstrating significant anti-inflammatory, anticancer, and anti-infective activities. It is a widely used non-steroidal anti-inflammatory drug (NSAID) suitable for various research applications.

  • Potent, orally active COX1/2 inhibitor
  • Exhibits anticancer activity
  • Exhibits anti-infective activity
  • Suitable for research applications in cancer, inflammation, and viral infection
  • Inhibits both COX-1 (IC50 18 nM) and COX-2 (IC50 26 nM)
  • Provides anti-inflammatory and analgesic effects
  • Used to prevent preterm labor due to prostaglandin synthesis inhibition
  • Used in premature infants with patent ductus arteriosus

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.