Learn More
Enzo Life Sciences Indomethacin (5g). CAS: 53-86-1
SDP
Supplier:  Enzo Life Sciences ALX270086G005
Encompass_Preferred
Non-steroidal anti-inflammatory and analgesic agent. Inhibits cyclooxygenase (IC50=0.1µM) selectively over lipoxygenases (IC50=100µM for 5-, 12- and 15-LO). Preferentially inhibits PGH synthase-1 over PGH synthase-2 (ID50=4.9-8.1 and 130-160µM respectively). Activates PPARα and γ receptors and induces adipocyte differentiation (EC50=8µM). A clinically useful NSAID. Formula: C19H16ClNO4. MW: 357.8. Purity: ≥98% (HPLC). Solubility: Soluble in acetone or methanol; insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-200-8401