Learn More
Cayman Chemical IndomethacIn-d4 500ug
SDP
Catalog No. 502935934
Encompass_Preferred
An internal standard for the quantification of indomethacin by GC- or LC-MS
Catalog No. 50-293-5934 Supplier Cayman Chemical Supplier No. 25434500 UG
An internal standard for the quantification of indomethacin by GC- or LC-MS