Learn More
Cayman Chemical IndomethacIn heptyl ester 50mg
SDP
Catalog No. 502894768
Encompass_Preferred
A potent, non-selective COX-2 inhibitor (IC50 = 0.04 µM); >1,700 times more potent as an inhibitor of COX-2 than COX-1
Catalog No. 50-289-4768 Supplier Cayman Chemical Supplier No. 7027150 MG
A potent, non-selective COX-2 inhibitor (IC50 = 0.04 µM); >1,700 times more potent as an inhibitor of COX-2 than COX-1