Learn More
Selleck Chemical LLC Indomethacin NSC-77541-50mg
SDP
Catalog No. 502389935
Encompass_Preferred
Indomethacin is a nonselective COX1 and COX2 inhibitor with IC50 of 0.1 μg/mL and 5 μg/mL, respectively, used to reduce fever, pain, stiffness, and swelling.
Catalog No. 50-238-9935 Supplier Selleck Chemical LLC Supplier No. S172350MG
Indomethacin is a nonselective COX1 and COX2 inhibitor with IC50 of 0.1 μg/mL and 5 μg/mL, respectively, used to reduce fever, pain, stiffness, and swelling.