Learn More
Apexbio Technology LLC Indomethacin-sodium-triH2O
SDP
Catalog No. 5000568783
Encompass_Preferred
Indomethacin sodium trihydrate (CAS No 74252-25-8) is a nonsteroidal anti-inflammatory drug (NSAID) used primarily in biomedical research It originates from indomethacin which works by inhibiting cyclooxygenase (COX) enzymes reducing prostaglandin synthesis and inflammation In vitro activity is observed in the nanomolar to low micromolar range highlighting its potency It is commonly applied in cell models animal studies and drug screening assays to investigate inflammatory processes Experimental concentrations typically depend on the specific research design ensuring adaptability to various scientific protocols
Catalog No. 50-005-68783 Supplier Apexbio Technology LLC Supplier No. C6491500
Indomethacin sodium trihydrate (CAS No 74252-25-8) is a nonsteroidal anti-inflammatory drug (NSAID) used primarily in biomedical research It originates from indomethacin which works by inhibiting cyclooxygenase (COX) enzymes reducing prostaglandin synthesis and inflammation In vitro activity is observed in the nanomolar to low micromolar range highlighting its potency It is commonly applied in cell models animal studies and drug screening assays to investigate inflammatory processes Experimental concentrations typically depend on the specific research design ensuring adaptability to various scientific protocols