Learn More
Medchemexpress LLC Indomethacin (Standard) | 53-86-1 | 99.6% | C₁₉H₁₆ClNO₄ | 250 MG
SDP
Catalog No. 5000352608
Encompass_Preferred
Indomethacin (Standard) is the analytical standard for Indomethacin, intended for research and analytical applications. It is a potent, orally active COX1/2 inhibitor, with IC50 values of 18 nM for COX-1 and 26 nM for COX-2. It exhibits both anticancer and anti-infective activities.
  • Analytical standard for Indomethacin.
  • Potent, orally active COX1/2 inhibitor.
  • IC50 values: 18 nM for COX-1 and 26 nM for COX-2.
  • Possesses anticancer activity.
  • Shows anti-infective activity.
  • Used in research for cancer, inflammation, and viral infections.
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-52608 Supplier Medchemexpress LLC Supplier No. 250MGHY14397R
Indomethacin (Standard) is the analytical standard for Indomethacin, intended for research and analytical applications. It is a potent, orally active COX1/2 inhibitor, with IC50 values of 18 nM for COX-1 and 26 nM for COX-2. It exhibits both anticancer and anti-infective activities.
  • Analytical standard for Indomethacin.
  • Potent, orally active COX1/2 inhibitor.
  • IC50 values: 18 nM for COX-1 and 26 nM for COX-2.
  • Possesses anticancer activity.
  • Shows anti-infective activity.
  • Used in research for cancer, inflammation, and viral infections.
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.