Learn More
Medchemexpress LLC Indomethacin (Standard) | 53-86-1 | 99.8% | C19H16ClNO4 | 25 MG
SDP
Catalog No. 5000356871
Encompass_Preferred
Indomethacin (Standard) serves as an analytical standard for Indomethacin, designed for various research and analytical applications. It is recognized as a potent, orally active COX1/2 inhibitor, demonstrating IC50 values of 18 nM for COX-1 and 26 nM for COX-2. This compound also exhibits significant anticancer and anti-infective properties, making it valuable for research focused on cancer, inflammation, and viral infections.
  • Analytical standard for Indomethacin
  • Intended for research and analytical applications
  • Potent, orally active COX1/2 inhibitor
  • IC50 values of 18 nM for COX-1 and 26 nM for COX-2
  • Exhibits anticancer activities
  • Exhibits anti-infective activities
  • Utilized for research in cancer, inflammation, and viral infections
Catalog No. 50-003-56871 Supplier Medchemexpress LLC Supplier No. 25MGHY14397R
Indomethacin (Standard) serves as an analytical standard for Indomethacin, designed for various research and analytical applications. It is recognized as a potent, orally active COX1/2 inhibitor, demonstrating IC50 values of 18 nM for COX-1 and 26 nM for COX-2. This compound also exhibits significant anticancer and anti-infective properties, making it valuable for research focused on cancer, inflammation, and viral infections.
  • Analytical standard for Indomethacin
  • Intended for research and analytical applications
  • Potent, orally active COX1/2 inhibitor
  • IC50 values of 18 nM for COX-1 and 26 nM for COX-2
  • Exhibits anticancer activities
  • Exhibits anti-infective activities
  • Utilized for research in cancer, inflammation, and viral infections