Learn More
Medchemexpress LLC Indomethacin (Standard) | 53-86-1 | MFCD00057095 | 99.81% | C₁₉H₁₆ClNO₄ | 50 MG
SDP
Catalog No. 5000354263
Encompass_Preferred
Indomethacin (Standard) is an analytical standard of Indomethacin, intended for research and analytical applications. It is a potent, orally active COX1/2 inhibitor with IC50 values of 18 nM and 26 nM for COX-1 and COX-2, respectively. This compound exhibits anticancer and anti-infective activities and can be utilized in research related to cancer, inflammation, and viral infections. It is commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Indomethacin
  • Potent, orally active COX1/2 inhibitor
  • Exhibits anticancer activity
  • Possesses anti-infective activity
  • Useful for cancer, inflammation, and viral infection research
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-003-54263 Supplier Medchemexpress LLC Supplier No. HY14397R50MG
Indomethacin (Standard) is an analytical standard of Indomethacin, intended for research and analytical applications. It is a potent, orally active COX1/2 inhibitor with IC50 values of 18 nM and 26 nM for COX-1 and COX-2, respectively. This compound exhibits anticancer and anti-infective activities and can be utilized in research related to cancer, inflammation, and viral infections. It is commonly employed in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for Indomethacin
  • Potent, orally active COX1/2 inhibitor
  • Exhibits anticancer activity
  • Possesses anti-infective activity
  • Useful for cancer, inflammation, and viral infection research
  • Suitable for HPLC, GC, and MS research experiments