Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indophenol Blue, TCI America™
SDP

Catalog No. I02081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I02081G Supplier TCI America Supplier No. I02081G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 132-31-0
Molecular Formula C18H16N2O
Molecular Weight (g/mol) 276.339
MDL Number MFCD00041739
InChI Key VRZJGENLTNRAIG-UHFFFAOYSA-N
Synonym N-(4-Dimethylaminophenyl)-1,4-naphthoquinoneimine, alpha-Naphthol Blue
PubChem CID 67235
IUPAC Name 4-[4-(dimethylamino)phenyl]iminonaphthalen-1-one
SMILES CN(C)C1=CC=C(C=C1)N=C2C=CC(=O)C3=CC=CC=C23

Specifications

Color Black/Purple
Quantity 1 g
Physical Form Crystal/Powder
Formula Weight 276.34
Chemical Name or Material Indophenol Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.