Learn More
Medchemexpress LLC Indotecan hydrochloride | 1228035-68-4 | 99.8% | 514.95 | 5 MG
SDP
Catalog No. 5000387760
Encompass_Preferred
Indotecan hydrochloride, an indenoisoquinoline derivative, is a potent Topoisomerase I inhibitor. It prevents the relaxation of supercoiled DNA and can be used for the research of visceral leishmaniasis.
  • Potent Topoisomerase I inhibitor.
  • Can be used for the research of visceral leishmaniasis.
Catalog No. 50-003-87760 Supplier Medchemexpress LLC Supplier No. HY18351A5MG
Indotecan hydrochloride, an indenoisoquinoline derivative, is a potent Topoisomerase I inhibitor. It prevents the relaxation of supercoiled DNA and can be used for the research of visceral leishmaniasis.
  • Potent Topoisomerase I inhibitor.
  • Can be used for the research of visceral leishmaniasis.