Learn More
Medchemexpress LLC Indotecan (LMP-400) | 915303-09-2 | 98.0% | 478.49 | 1 MG
SDP
Catalog No. 5000387979
Encompass_Preferred
Indotecan (LMP-400), an indenoisoquinoline derivative, is a potent Topoisomerase I inhibitor, with IC50s of 300, 1200, 560 nM for P388, HCT116, MCF-7 cell lines, respectively. It prevents the relaxation of supercoiled DNA and can be used for the research of visceral leishmaniasis.
  • Potent Topoisomerase I inhibitor.
  • Effective against P388, HCT116, and MCF-7 cell lines with specified IC50 values.
  • Prevents relaxation of supercoiled DNA.
  • Applicable for research on visceral leishmaniasis.
  • Available in various quantities (1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg).
  • Available in other forms such as Indotecan hydrochloride.
Catalog No. 50-003-87979 Supplier Medchemexpress LLC Supplier No. HY183511MG
Indotecan (LMP-400), an indenoisoquinoline derivative, is a potent Topoisomerase I inhibitor, with IC50s of 300, 1200, 560 nM for P388, HCT116, MCF-7 cell lines, respectively. It prevents the relaxation of supercoiled DNA and can be used for the research of visceral leishmaniasis.
  • Potent Topoisomerase I inhibitor.
  • Effective against P388, HCT116, and MCF-7 cell lines with specified IC50 values.
  • Prevents relaxation of supercoiled DNA.
  • Applicable for research on visceral leishmaniasis.
  • Available in various quantities (1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg).
  • Available in other forms such as Indotecan hydrochloride.