Learn More
Cayman Chemical (±)-Indoxacarb, Purity: ≥95%, CAS: 144171-61-9, 100 mg
SDP
Catalog No. 502938057
Encompass_Preferred
A broad-spectrum insecticide; topical adminstration is lethal to various insects (LD50s = 0.03->7.8 ng/mg); suppresses voltage-gated sodium channel currents in rat dorsal root ganglion neurons in a concentration-dependent manner
Catalog No. 50-293-8057 Supplier Cayman Chemical Supplier No. 29165100 MG
A broad-spectrum insecticide; topical adminstration is lethal to various insects (LD50s = 0.03->7.8 ng/mg); suppresses voltage-gated sodium channel currents in rat dorsal root ganglion neurons in a concentration-dependent manner