Learn More
Chem Service Inc Indoxacarb Solution, 144171-61-9, MFCD03792834, 1 mL
SDP
Catalog No. NC1529045
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Molecular Formula: C22 H17 Cl F3 N3 O7, Molecular Weight: 527.837, Concentration and Solvent: 100ug/mL in Acetonitrile.
Catalog No. NC1529045 Supplier Chem Service Inc Supplier No. S12895A11ML
This item has a minimum order quantity of 2 per supplier requirements
Molecular Formula: C22 H17 Cl F3 N3 O7, Molecular Weight: 527.837, Concentration and Solvent: 100ug/mL in Acetonitrile.