Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indoximod | 2071683-99-1 | 98.8% | 395.92 g/mol | C20H30ClN3O3 | 5 MG
SDP

Catalog No. 5000221149
Encompass_Preferred

NLG802 is an experimental prodrug of indoximod designed to act as an orally active indoleamine 2,3-dioxygenase (IDO) inhibitor for research applications. It is supplied as a white to off-white solid and is available in solid and solution formats suitable for in-vitro and in-vivo studies.

  • Prodrug of indoximod, an orally active IDO inhibitor.
  • High purity (≈98.8%).
  • Supplied as a white to off-white solid.
  • Available in small solid quantities and 10 mM DMSO solutions.
  • Soluble in DMSO at 250 mg/mL; ultrasonic assistance recommended.
  • Storage: solid at 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-21149 Supplier Medchemexpress LLC Supplier No. HY1453465MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NLG802 is an experimental prodrug of indoximod designed to act as an orally active indoleamine 2,3-dioxygenase (IDO) inhibitor for research applications. It is supplied as a white to off-white solid and is available in solid and solution formats suitable for in-vitro and in-vivo studies.

  • Prodrug of indoximod, an orally active IDO inhibitor.
  • High purity (≈98.8%).
  • Supplied as a white to off-white solid.
  • Available in small solid quantities and 10 mM DMSO solutions.
  • Soluble in DMSO at 250 mg/mL; ultrasonic assistance recommended.
  • Storage: solid at 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.