Learn More
Selleck Chemical LLC Indoximod
SDP
Catalog No. 5000004558
Encompass_Preferred
Indoximod a methylated tryptophan acts as an IDO (indoleamine-(2 3)-dioxygenase) pathway inhibitor and reverses IDO-mediated immune suppression Phase 2 This product has poor solubility animal experiments are available cell experiments please choose carefully
Catalog No. 50-000-04558 Supplier Selleck Chemical LLC Supplier No. S7756200MG
Indoximod a methylated tryptophan acts as an IDO (indoleamine-(2 3)-dioxygenase) pathway inhibitor and reverses IDO-mediated immune suppression Phase 2 This product has poor solubility animal experiments are available cell experiments please choose carefully