Learn More
Abcam Indo x imod, Indoleamine 2,3-dio x ygenase (IDO) inhibitor, 500MG
Catalog No. 502672643
Encompass_Preferred
MW 218.25 Da, Purity >95%. Methylated tryptophan which acts as an Indoleamine 2,3-dioxygenase (IDO) inhibitor. Prevents T-cell anergy triggered by dendritic cells overexpressing IDO in mice.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2643 Supplier Abcam Supplier No. AB231506500MG
MW 218.25 Da, Purity >95%. Methylated tryptophan which acts as an Indoleamine 2,3-dioxygenase (IDO) inhibitor. Prevents T-cell anergy triggered by dendritic cells overexpressing IDO in mice.
The product is subject to the following: Abcam Restricted Use Statement