Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indoxyl beta-D-Glucoside, Ultrapure, 97%, Spectrum™ Chemical
SDP

Catalog No. 18611031 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-031 50 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-611-031 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I326250GMBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 487-60-5
Molecular Weight (g/mol) 295.29

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 50 g
Percent Purity 0.97
Grade Ultra Pure
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Indoxyl beta-D-Glucoside

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.