Learn More
Abcam Indo x yl -Glucuronide, Chromogenic beta-D-glucuronidase substrate, 100MG
Catalog No. 502704672
Encompass_Preferred
MW 408.5 g/mol, Purity >99%. Chromogenic substrate for β-D-glucuronidase, employed in the detection and enumeration of E.coli. Yields a blue precipitate upon cleavage.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4672 Supplier Abcam Supplier No. AB146380100MG
MW 408.5 g/mol, Purity >99%. Chromogenic substrate for β-D-glucuronidase, employed in the detection and enumeration of E.coli. Yields a blue precipitate upon cleavage.
The product is subject to the following: Abcam Restricted Use Statement