Learn More
Cayman Chemical Indoxyl Sulfate-d5 (potassium salt), Purity: ≥99% deuterated forms (d1-d5), CAS: 1644451-34-2, 10 mg
SDP
Catalog No. 502950487
Encompass_Preferred
An internal standard for the quantification of indoxyl sulfate by GC- or LC-MS
Catalog No. 50-295-0487 Supplier Cayman Chemical Supplier No. 3451110 MG
An internal standard for the quantification of indoxyl sulfate by GC- or LC-MS