Learn More
Cayman Chemical Indoxyl Sulfate (potassium salt), Purity: ≥98%, CAS: 2642-37-7, 1 g
SDP
Catalog No. 502966587
Encompass_Preferred
A uremic toxin and metabolite of tryptophan; activates AhR in HepG2 40/6 hepatoma cells (EC50 = 12.1 nM in a reporter assay); inhibits OAT1 and OAT3 (Kis = 34.2 and 74.4 µM, respectively for the rat transporters) in S2 proximal tubule cells; increases superoxide anion and nitric oxide levels in isolated human mononuclear blood cells at 0.2 and 1 mM; increases serum creatinine and BUN levels in the 5/6 nephrectomized rat model of chronic renal failure at 50 mg/kg
Catalog No. 50-296-6587 Supplier Cayman Chemical Supplier No. 169261 G
A uremic toxin and metabolite of tryptophan; activates AhR in HepG2 40/6 hepatoma cells (EC50 = 12.1 nM in a reporter assay); inhibits OAT1 and OAT3 (Kis = 34.2 and 74.4 µM, respectively for the rat transporters) in S2 proximal tubule cells; increases superoxide anion and nitric oxide levels in isolated human mononuclear blood cells at 0.2 and 1 mM; increases serum creatinine and BUN levels in the 5/6 nephrectomized rat model of chronic renal failure at 50 mg/kg