Learn More
Grainger Induced Draft Furnace Blower 115V
Catalog No. 502749132
Encompass_Preferred
The Grainger Approved Induced Draft Furnace Blower is a 1/3 HP, 115V, 60Hz, 1-phase, 3450 RPM blower motor designed for residential and light commercial furnaces. It features sleeve bearings and manual reset thermal overload protection. The blower has a shipped weight of 9 lbs and dimensions of 11-1/4" length x 8-3/4" width x 8-1/2" height. This furnace blower can provide airflow up to 1300 CFM and static pressure up to 0.8 inches water gauge. The motor is thermally protected for safety.
Catalog No. 50-274-9132 Supplier Grainger Supplier No. 14C434
The Grainger Approved Induced Draft Furnace Blower is a 1/3 HP, 115V, 60Hz, 1-phase, 3450 RPM blower motor designed for residential and light commercial furnaces. It features sleeve bearings and manual reset thermal overload protection. The blower has a shipped weight of 9 lbs and dimensions of 11-1/4" length x 8-3/4" width x 8-1/2" height. This furnace blower can provide airflow up to 1300 CFM and static pressure up to 0.8 inches water gauge. The motor is thermally protected for safety.