Learn More
Grainger Induced Draft Furnace Blower 115V
Catalog No. 502749049
Encompass_Preferred
The Packard 14C433 is an induced draft furnace blower designed for use with oil or gas fired heating systems. It features a 1/3 HP motor with permanently lubricated ball bearings and automatic reset thermal overload protection. The blower has a listed airflow of 850 CFM and develops 0.5 inches static pressure at high speed. It measures 14-1/2 x 14-1/2 x 8 inches and has a 6 inch diameter outlet. This UL listed blower can be used as a retrofit or replacement blower for many furnace applications.
Catalog No. 50-274-9049 Supplier Grainger Supplier No. 14C433
The Packard 14C433 is an induced draft furnace blower designed for use with oil or gas fired heating systems. It features a 1/3 HP motor with permanently lubricated ball bearings and automatic reset thermal overload protection. The blower has a listed airflow of 850 CFM and develops 0.5 inches static pressure at high speed. It measures 14-1/2 x 14-1/2 x 8 inches and has a 6 inch diameter outlet. This UL listed blower can be used as a retrofit or replacement blower for many furnace applications.