Learn More
Grainger Induced Draft Furnace Blower 115V
Catalog No. 502749067
Encompass_Preferred
The Packard 14C434 is an induced draft furnace blower designed for use with oil or gas fired heating systems. It features a 1/3 HP PSC motor with permanently lubricated ball bearings and automatic reset thermal overload protection. The blower has a listed airflow of 850 CFM and develops 0.5 inches static pressure at high speed. It operates on 115V power and measures 14-1/2 x 14-1/2 x 8 inches. This UL listed blower motor is ETL certified to meet CSA standards. The 14C434 has a 1 year warranty and is made in the USA.
Catalog No. 50-274-9067 Supplier Grainger Supplier No. 14C435
The Packard 14C434 is an induced draft furnace blower designed for use with oil or gas fired heating systems. It features a 1/3 HP PSC motor with permanently lubricated ball bearings and automatic reset thermal overload protection. The blower has a listed airflow of 850 CFM and develops 0.5 inches static pressure at high speed. It operates on 115V power and measures 14-1/2 x 14-1/2 x 8 inches. This UL listed blower motor is ETL certified to meet CSA standards. The 14C434 has a 1 year warranty and is made in the USA.