Learn More
Grainger Induced Draft Furnace Blower 115V
Catalog No. 502748990
Encompass_Preferred
The Packard 14C432 is an induced draft furnace blower designed for use with oil or gas fired heating systems. It features a 1/3 HP motor with permanently lubricated ball bearings and thermal overload protection. The blower wheel is made of steel with forward curved blades. It has a rated airflow of 800 CFM at 0.5 inches static pressure. The housing is constructed of heavy gauge steel and has 6 inch inlet and outlet openings. This blower can provide the airflow needed for residential or light commercial heating systems. Its rugged construction makes it a durable choice. With proper installation and maintenance, this Packard blower should provide years of reliable performance.
Catalog No. 50-274-8990 Supplier Grainger Supplier No. 14C432
The Packard 14C432 is an induced draft furnace blower designed for use with oil or gas fired heating systems. It features a 1/3 HP motor with permanently lubricated ball bearings and thermal overload protection. The blower wheel is made of steel with forward curved blades. It has a rated airflow of 800 CFM at 0.5 inches static pressure. The housing is constructed of heavy gauge steel and has 6 inch inlet and outlet openings. This blower can provide the airflow needed for residential or light commercial heating systems. Its rugged construction makes it a durable choice. With proper installation and maintenance, this Packard blower should provide years of reliable performance.