Learn More
IPHASE PHARMA SERVICES LLC Induced Golden Syrian Hamster Liver S9,Male,35mg/mL,1mL
Catalog No. NC3966972
Encompass_Preferred
Induced Hamster (Golden Syrian) liver S9 applied on Enhanced Ames test, to assess the carcinogenic potential of n-nitrosamines.
Catalog No. NC3966972 Supplier IPHASE PHARMA SERVICES LLC Supplier No. 027F1041
Induced Hamster (Golden Syrian) liver S9 applied on Enhanced Ames test, to assess the carcinogenic potential of n-nitrosamines.