Learn More
IPHASE PHARMA SERVICES LLC Induced Sprague-Dawley Rat Liver S9,Male,35mg/mL,1mL
Catalog No. NC3966971
Encompass_Preferred
Induced rat liver S9 is a key component in assessing the genetic toxicity potential of chemicals. It is commonly used in combination with in vitro assays, such as the Ames test and mutation tests, to evaluate the mutagenic properties of compounds
Catalog No. NC3966971 Supplier IPHASE PHARMA SERVICES LLC Supplier No. 0271041
Induced rat liver S9 is a key component in assessing the genetic toxicity potential of chemicals. It is commonly used in combination with in vitro assays, such as the Ames test and mutation tests, to evaluate the mutagenic properties of compounds