Learn More
Medchemexpress LLC Inducible T-cell co-stimulator (ICOS), rat, HEK293 Fc-fusion | >95.0% | 10 UG
SDP
Catalog No. 5000291655
Encompass_Preferred
Recombinant rat Inducible T-cell co-stimulator (ICOS) extracellular domain expressed in HEK293 cells as a C-terminal human Fc fusion. Supplied lyophilized and purified (>95%) from PBS, pH 7.4. Designed for in vitro immunology research including T-cell activation, receptor-ligand interaction assays, and biochemical characterization. For research use only.
  • HEK293-expressed recombinant rat ICOS extracellular domain.
  • C-terminal human Fc fusion for improved stability and detection.
  • Lyophilized formulation from PBS, pH 7.4.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Available in microgram-scale quantities for assay development.
Catalog No. 50-002-91655 Supplier Medchemexpress LLC Supplier No. HYP7486810UG
Add to Cart
Add to Cart
Recombinant rat Inducible T-cell co-stimulator (ICOS) extracellular domain expressed in HEK293 cells as a C-terminal human Fc fusion. Supplied lyophilized and purified (>95%) from PBS, pH 7.4. Designed for in vitro immunology research including T-cell activation, receptor-ligand interaction assays, and biochemical characterization. For research use only.
  • HEK293-expressed recombinant rat ICOS extracellular domain.
  • C-terminal human Fc fusion for improved stability and detection.
  • Lyophilized formulation from PBS, pH 7.4.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Available in microgram-scale quantities for assay development.