Learn More
Hach Company Induct Electrodeless Sens-San
Catalog No. 5000207649 Shop All Hach Company Products
Encompass
Sanitary Inductive-Electrodeless Conductivity Sensor-PP Body MaterialPolypropylene Sanitary (CIP) Flange Mounting Style 6m (20 ft) Analog Cable Electrodeless Conductivity Sensor
Catalog No. 50-002-07649 Supplier Hach Company Supplier No. 3705E2T
Sanitary Inductive-Electrodeless Conductivity Sensor-PP Body MaterialPolypropylene Sanitary (CIP) Flange Mounting Style 6m (20 ft) Analog Cable Electrodeless Conductivity Sensor