Learn More
Stoelting™ Induction Chamber
SDP
Catalog No. 10000630 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Rat
Catalog No. Animal
10-000-630 Rat
1 options
Catalog No. 10-000-630 Supplier Stoelting™ Supplier No. 50216
Add to Cart
Add to Cart
Stoelting™ Induction Chamber is for the initial anesthetization of animals difficult to keep masked when awake.
Route gas through chamber, then to mask or 50206 Scavenger. 
Note:  Rubber tubing is not included; sold separately.
Specifications
For Use With (Application) Neuroscience
Animal Rat
Includes Induction Chamber, inlet port connector. (Tubing not included.)
Type Induction Chamber