Learn More
Vetequip Inc INDUCTION CHAMBER 2L
SDP
Catalog No. NC9905720
Encompass_Preferred
Induction Chamber, 2L, 9W x 3 3/4 D x 3 3/4 in. H, Designed for one mouse or a cage full, or a large rat up to 650g, Large easy-to-use latches, Positive sealing gasket at lid helps prevent leakage, Durable, Clear acrylic
Catalog No. NC9905720 Supplier Vetequip Inc Supplier No. 941444
Induction Chamber, 2L, 9W x 3 3/4 D x 3 3/4 in. H, Designed for one mouse or a cage full, or a large rat up to 650g, Large easy-to-use latches, Positive sealing gasket at lid helps prevent leakage, Durable, Clear acrylic