Learn More
Stoelting™ Induction Chamber for Active Gas Scavenger
SDP
Catalog No. 10000932 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-932 Neuroscience
1 options
Catalog No. 10-000-932 Supplier Stoelting™ Supplier No. 53915
Add to Cart
Add to Cart
Induction Chamber for Active Gas Scavenger works with active vacuum waste gas management systems.
Features a slide top style lid which reduces exposure to waste anesthetic gases compared to lift style lid induction chambers.
Specifications
For Use With (Application) Neuroscience
For Use With (Equipment) Active Gas Scavenger
Includes Slide Top