Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOMNI Scientific™ Medium Induction Chamber Clear, Active Style
SDP

Catalog No. 01670804 Shop All Somni Scientific Products
Change view
Click to view available options
Includes:
Active Style
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Size
01-670-804 Active Style Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-804 Supplier SOMNI Scientific™ Supplier No. IC3014A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The SOMNI Medium Active Induction Chamber is constructed of durable, shatter resistant polycarbonate. The slide top lid keeps anesthetic gases from being drawn into the workplace. Convenient one-handed operation. The chamber is supplied with color coded inlet and outlet ports.

  • Active style prevents unnecessary exposure when used with active waste gas removal system
  • Slide top lid
  • One-handed operation
  • Color coded inlet and outlet ports
  • Compatible with 19 mm tubing and 15 mm adapters
  • Can be cleaned with common clinical disinfectants

Specifications

For Use With (Application) Animal Inhalant Anesthesia
Size Medium
Animal Mouse
Includes Active Style
Type Induction Chamber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.