Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOMNI Scientific™ Medium Induction Chamber Clear, Passive Style
SDP

Catalog No. 01670805 Shop All Somni Scientific Products
Change view
Click to view available options
Includes:
Passive Style
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Size
01-670-805 Passive Style Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-805 Supplier SOMNI Scientific™ Supplier No. IC3014P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The SOMNI Medium Passive Induction Chamber is constructed of durable, shatter resistant polycarbonate. The slide top lid keeps anesthetic gases from being drawn into the workplace. Convenient one-handed operation. The chamber is supplied with color coded inlet and outlet ports

  • Slide top lid keeps anesthetic from being drawn into workplace
  • Constructed of shatter resistant polycarbonate
  • One-handed operation
  • Color coded inlet and outlet ports
  • Compatible with 19 mm tubing and 15 mm adapters

Specifications

For Use With (Application) Animal Inhalant Anesthesia
Size Medium
Animal Mouse
Includes Passive Style
Type Induction Chamber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.