Learn More
Euthanex Sure-Seal Guinea Pig/Rabbit Chamber, 18"" L x 12"" W x 12"" H.
Catalog No. 501540172
Encompass_Preferred
Sure-Seal Guinea Pig/Rabbit Chamber, 18" L x 12" W x 12" H, heavy-duty acrylic, gasket seal, locking latch. Gas is exhausted through an outlet port that is tube-connected to a charcoal filter canister. Accommodates oxygen-flush before transferring animals out of chamber.
Catalog No. 50-154-0172 Supplier Euthanex Supplier No. EZ179
Sure-Seal Guinea Pig/Rabbit Chamber, 18" L x 12" W x 12" H, heavy-duty acrylic, gasket seal, locking latch. Gas is exhausted through an outlet port that is tube-connected to a charcoal filter canister. Accommodates oxygen-flush before transferring animals out of chamber.