Learn More
Hach Company Inductive Conduc. sensor Peek
Catalog No. 5000207777 Shop All Hach Company Products
Encompass
Digital Convertible Mounting Style Inductive (Electrodeless) Conductivity Sensor with Peek Body MaterialPEEK Convertible Mounting Style 6m (20ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable
Catalog No. 50-002-07777 Supplier Hach Company Supplier No. D3727E2T
Digital Convertible Mounting Style Inductive (Electrodeless) Conductivity Sensor with Peek Body MaterialPEEK Convertible Mounting Style 6m (20ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable