Learn More
Hach Company Inductive Conduc. sensor PFA
Catalog No. 5000207778 Shop All Hach Company Products
Encompass
Digital Convertible Mounting Style Inductive-Electrodeless Conductivity Sensor-PFA Body MaterialPFA Convertible Mounting Style 6m (20 ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable
Catalog No. 50-002-07778 Supplier Hach Company Supplier No. D3728E2T
Digital Convertible Mounting Style Inductive-Electrodeless Conductivity Sensor-PFA Body MaterialPFA Convertible Mounting Style 6m (20 ft) Analog Cable Electrodeless Conductivity Sensor includes 6120800 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable